Gene Transcripts

Gene Symbol
4930579K19Rik
Entrez Gene ID
75881
Gene Name
RIKEN cDNA 4930579K19 gene
Gene Aliases
Chromosome Location
Chr. 9: 98562460 - 98563611 on Build GRCm38
UniGene ID
Mm.483402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK016329.1 - 1 488
AK077474.1 - 1 498
AK087620.1 - 1 517
BC092140.1 - 1 457

View Full Product Details