Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177471.4 | NP_803422.2 | 3 | 235 | |
GenBank mRNA | AK008760.1 | - | 3 | 206 | |
AK079810.1 | - | 3 | 236 | ||
AK135942.1 | - | 3 | 212 | ||
AK170686.1 | - | 3 | 185 | ||
BC096763.1 | - | 3 | 193 | 30053993 |