gRNA Design Details

Target DNA Sequence
GCCAAAAAGTATAAACTTTG
PAM Sequence
GGG
Target locus
Chr.7: 23729184 - 23729206 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 78.94/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK31
Entrez Gene ID
56164
Gene Name
serine/threonine kinase 31
Gene Aliases
SGK396, TDRD8
Chromosome Location
Chr.7: 23710141 - 23832513 on GRCh38
UniGene ID
Hs.309767
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001260504.1 NP_001247433.1 6
NM_001260505.1 NP_001247434.1 6
NM_031414.4 NP_113602.2 6
NM_032944.3 NP_116562.2 6
NR_048542.1 6
XM_006715758.3 XP_006715821.1 6
XM_011515449.2 XP_011513751.2 6
XM_011515450.1 XP_011513752.1 6
XM_011515452.2 XP_011513754.2 6