gRNA Design Details
- Target DNA Sequence
- TTCCGATGTAGCCACTGCCC
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 69642932 - 69642954 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.76/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AAK1
- Entrez Gene ID
-
22848
- Gene Name
- AP2 associated kinase 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 69457995 - 69643845 on GRCh38
- UniGene ID
- Hs.468878
- Species
-
Human