gRNA Design Details
- Target DNA Sequence
- GACCTCCTTGGCGTAGTAGT
- PAM Sequence
- CGG
- Target locus
-
Chr.19: 41352721 - 41352743 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFB1
- Entrez Gene ID
-
7040
- Gene Name
- transforming growth factor beta 1
- Gene Aliases
-
CED, DPD1, LAP, TGFB, TGFbeta
- Chromosome Location
-
Chr.19: 41330531 - 41353933 on GRCh38
- UniGene ID
- Hs.645227
- Species
-
Human