gRNA Design Details
- Target DNA Sequence
- GTACCCTTCCAGGAAGTGTG
- PAM Sequence
- GGG
- Target locus
-
Chr.14: 74949529 - 74949551 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.81/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PGF
- Entrez Gene ID
-
5228
- Gene Name
- placental growth factor
- Gene Aliases
-
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760
- Chromosome Location
-
Chr.14: 74941830 - 74955764 on GRCh38
- UniGene ID
- Hs.252820
- Species
-
Human