gRNA Design Details

Target DNA Sequence
ATCCATGAAGCTAATGGCTG
PAM Sequence
AGG
Target locus
Chr.15: 83572590 - 83572612 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.53/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SH3GL3
Entrez Gene ID
6457
Gene Name
SH3 domain containing GRB2 like 3, endophilin A3
Gene Aliases
CNSA3, EEN-B2, HsT19371, SH3D2C, SH3P13
Chromosome Location
Chr.15: 83447123 - 83618743 on GRCh38
UniGene ID
Hs.270055
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001301108.1 NP_001288037.1 5
NM_001301109.1 NP_001288038.1 8
NM_001324182.1 NP_001311111.1 5
NM_001324183.1 NP_001311112.1 6
NM_001324184.1 NP_001311113.1 7
NM_001324185.1 NP_001311114.1 6
NM_001324186.1 NP_001311115.1 6
NM_001324187.1 NP_001311116.1 5
NM_003027.4 NP_003018.3 5
NR_125370.1 5
NR_136711.1 6
NR_136712.1 6
NR_136713.1 5
NR_136714.1 5
XM_011521889.1 XP_011520191.1 7
XM_011521891.2 XP_011520193.1 6
XM_011521892.2 XP_011520194.1 6
XM_017022486.1 XP_016877975.1 5