gRNA Design Details

Target DNA Sequence
GAACTTACTCATGGGCAAGG
PAM Sequence
AGG
Target locus
Chr.22: 39960091 - 39960113 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 88.83/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GRAP2
Entrez Gene ID
9402
Gene Name
GRB2-related adaptor protein 2
Gene Aliases
GADS, GRAP-2, GRB2L, GRBLG, GRID, GRPL, GrbX, Grf40, Mona, P38
Chromosome Location
Chr.22: 39901073 - 39973343 on GRCh38
UniGene ID
Hs.517499
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001291824.1 NP_001278753.1 4
NM_001291825.1 NP_001278754.1 3
NM_001291826.1 NP_001278755.1 3
NM_001291828.1 NP_001278757.1 2
NM_004810.3 NP_004801.1 4
XM_006724376.3 XP_006724439.1 3