gRNA Design Details
- Target DNA Sequence
- GTATGACTCCCGGCTGAACA
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 35875566 - 35875588 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCL5
- Entrez Gene ID
-
6352
- Gene Name
- C-C motif chemokine ligand 5
- Gene Aliases
-
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
- Chromosome Location
-
Chr.17: 35871491 - 35880373 on GRCh38
- UniGene ID
- Hs.514821
- Species
-
Human