gRNA Design Details
- Target DNA Sequence
- AACAAGCGTCCAAAAGAACC
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 17589491 - 17589513 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PDGFRL
- Entrez Gene ID
-
5157
- Gene Name
- platelet derived growth factor receptor like
- Gene Aliases
-
PDGRL, PRLTS
- Chromosome Location
-
Chr.8: 17576433 - 17643133 on GRCh38
- UniGene ID
- Hs.458573
- Species
-
Human