gRNA Design Details

Target DNA Sequence
GGCATCATACTCCATCTGCA
PAM Sequence
GGG
Target locus
Chr.1: 204469695 - 204469717 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.94/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PIK3C2B
Entrez Gene ID
5287
Gene Name
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 beta
Gene Aliases
C2-PI3K
Chromosome Location
Chr.1: 204422630 - 204494815 on GRCh38
UniGene ID
Hs.497487
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002646.3 NP_002637.3 3
XM_005245257.2 XP_005245314.1 2
XM_005245258.3 XP_005245315.1 2
XM_011509630.2 XP_011507932.1 4
XM_011509631.2 XP_011507933.1 3
XM_011509633.2 XP_011507935.1 2
XM_017001473.1 XP_016856962.1 3
XM_017001474.1 XP_016856963.1 3