gRNA Design Details
- Target DNA Sequence
- AGTTGCTCATGCAGGATTTC
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 30644841 - 30644863 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFBR2
- Entrez Gene ID
-
7048
- Gene Name
- transforming growth factor beta receptor 2
- Gene Aliases
-
AAT3, FAA3, LDS1B, LDS2, LDS2B, MFS2, RIIC, TAAD2, TGFR-2, TGFbeta-RII
- Chromosome Location
-
Chr.3: 30606490 - 30694142 on GRCh38
- UniGene ID
- Hs.604277
- Species
-
Human