gRNA Design Details
- Target DNA Sequence
- AGAGAGCACGGAGATAACTG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 45890570 - 45890592 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP1
- Entrez Gene ID
-
3484
- Gene Name
- insulin like growth factor binding protein 1
- Gene Aliases
-
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1
- Chromosome Location
-
Chr.7: 45888360 - 45893668 on GRCh38
- UniGene ID
- Hs.642938
- Species
-
Human