gRNA Design Details
- Target DNA Sequence
- GAGCACGGAGATAACTGAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 45890573 - 45890595 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.26/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP1
- Entrez Gene ID
-
3484
- Gene Name
- insulin like growth factor binding protein 1
- Gene Aliases
-
AFBP, IBP1, IGF-BP25, PP12, hIGFBP-1
- Chromosome Location
-
Chr.7: 45888360 - 45893668 on GRCh38
- UniGene ID
- Hs.642938
- Species
-
Human