gRNA Design Details
- Target DNA Sequence
- AGAGATGAAATGGACTTGTT
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 59659358 - 59659380 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
77.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PATL1
- Entrez Gene ID
-
219988
- Gene Name
- PAT1 homolog 1, processing body mRNA decay factor
- Gene Aliases
-
Pat1b, hPat1b
- Chromosome Location
-
Chr.11: 59636716 - 59669040 on GRCh38
- UniGene ID
- Hs.591960
- Species
-
Human