gRNA Design Details
- Target DNA Sequence
- CAGCAGTGCACGGACAACTT
- PAM Sequence
- TGG
- Target locus
-
Chr.18: 59469553 - 59469575 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCBE1
- Entrez Gene ID
-
147372
- Gene Name
- collagen and calcium binding EGF domains 1
- Gene Aliases
-
HKLLS1
- Chromosome Location
-
Chr.18: 59430939 - 59698181 on GRCh38
- UniGene ID
- Hs.34333
- Species
-
Human