gRNA Design Details

Target DNA Sequence
TGTCTCTGATTCGCATGTAA
PAM Sequence
AGG
Target locus
Chr.20: 13072303 - 13072325 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.85/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPTLC3
Entrez Gene ID
55304
Gene Name
serine palmitoyltransferase long chain base subunit 3
Gene Aliases
C20orf38, LCB2B, LCB3, SPT3, SPTLC2L, dJ718P11, dJ718P11.1, hLCB2b
Chromosome Location
Chr.20: 13008979 - 13169103 on GRCh38
UniGene ID
Hs.425023
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_018327.2 NP_060797.2 3
XM_011529279.1 XP_011527581.1 3
XM_017027922.1 XP_016883411.1 3