gRNA Design Details
- Target DNA Sequence
- GGAAGCACCCCTTCATGTTA
- PAM Sequence
- TGG
- Target locus
-
Chr.20: 13048995 - 13049017 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPTLC3
- Entrez Gene ID
-
55304
- Gene Name
- serine palmitoyltransferase long chain base subunit 3
- Gene Aliases
-
C20orf38, LCB2B, LCB3, SPT3, SPTLC2L, dJ718P11, dJ718P11.1, hLCB2b
- Chromosome Location
-
Chr.20: 13008979 - 13169103 on GRCh38
- UniGene ID
- Hs.425023
- Species
-
Human