gRNA Design Details
- Target DNA Sequence
- GAGCTCCGTCTACAAGCTTG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 45290616 - 45290638 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.07/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MAP3K14
- Entrez Gene ID
-
9020
- Gene Name
- mitogen-activated protein kinase kinase kinase 14
- Gene Aliases
-
FTDCR1B, HS, HSNIK, NIK
- Chromosome Location
-
Chr.17: 45263119 - 45317064 on GRCh38
- UniGene ID
- Hs.404183
- Species
-
Human