gRNA Design Details
- Target DNA Sequence
- GCTGACACACTTGCAGGCGT
- PAM Sequence
- TGG
- Application
- Gene Knock-Out
Score:
88.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF13
- Entrez Gene ID
-
2258
- Gene Name
- fibroblast growth factor 13
- Gene Aliases
-
FGF-13, FGF2, FHF-2, FHF2
- Chromosome Location
-
Chr.X: 138631573 - 139222889 on GRCh38
- UniGene ID
- Hs.6540
- Species
-
Human