gRNA Design Details
- Target DNA Sequence
- CTCATTCTCGTCAAACACGG
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 5051889 - 5051911 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK32B
- Entrez Gene ID
-
55351
- Gene Name
- serine/threonine kinase 32B
- Gene Aliases
-
HSA250839, STK32, STKG6, YANK2
- Chromosome Location
-
Chr.4: 5051323 - 5501001 on GRCh38
- UniGene ID
- Hs.133062
- Species
-
Human