gRNA Design Details

Target DNA Sequence
TTCTGCGGGCCATTGGTAAA
PAM Sequence
GGG
Target locus
Chr.4: 5139926 - 5139948 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK32B
Entrez Gene ID
55351
Gene Name
serine/threonine kinase 32B
Gene Aliases
HSA250839, STK32, STKG6, YANK2
Chromosome Location
Chr.4: 5051323 - 5501001 on GRCh38
UniGene ID
Hs.133062
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001306082.1 NP_001293011.1 2
NM_018401.2 NP_060871.1 2
XM_011513497.1 XP_011511799.1 2
XM_017008374.1 XP_016863863.1 1
XM_017008375.1 XP_016863864.1 1
XM_017008376.1 XP_016863865.1 1