gRNA Design Details
- Target DNA Sequence
- GGTGTAGCACCAGCTCACTC
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 47699565 - 47699587 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.52/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Frs3
- Entrez Gene ID
-
107971
- Gene Name
- fibroblast growth factor receptor substrate 3
- Gene Aliases
-
4930417B13Rik, AI449674, Frs2beta, Snt2
- Chromosome Location
-
Chr.17: 47687296 - 47704286 on GRCm38
- UniGene ID
- Mm.392206
- Species
-
Mouse