gRNA Design Details
- Target DNA Sequence
- ATTCAAGTGTTCGAGGGCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 47701633 - 47701655 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Frs3
- Entrez Gene ID
-
107971
- Gene Name
- fibroblast growth factor receptor substrate 3
- Gene Aliases
-
4930417B13Rik, AI449674, Frs2beta, Snt2
- Chromosome Location
-
Chr.17: 47687296 - 47704286 on GRCm38
- UniGene ID
- Mm.392206
- Species
-
Mouse