gRNA Design Details
- Target DNA Sequence
- TTGTACGTCCAGCTGGTGCA
- PAM Sequence
- GGG
- Application
- Gene Knockout
Score:
89.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FIGF
- Entrez Gene ID
-
2277
- Gene Name
- c-fos induced growth factor
- Gene Aliases
-
VEGF-D, VEGFD
- Chromosome Location
-
Chr.X: 15345591 - 15384413 on GRCh38
- UniGene ID
- Hs.11392
- Species
-
Human