gRNA Design Details
- Target DNA Sequence
- TCAGTTCTAGGAAAGCAGAA
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 24648981 - 24649003 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
84.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRRM1
- Entrez Gene ID
-
10250
- Gene Name
- serine and arginine repetitive matrix 1
- Gene Aliases
-
160-KD, POP101, SRM160
- Chromosome Location
-
Chr.1: 24643012 - 24673281 on GRCh38
- UniGene ID
- Hs.18192
- Species
-
Human