gRNA Design Details
- Target DNA Sequence
- GAGAATTTATGGGAGAACTG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 24648920 - 24648942 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
79.14/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRRM1
- Entrez Gene ID
-
10250
- Gene Name
- serine and arginine repetitive matrix 1
- Gene Aliases
-
160-KD, POP101, SRM160
- Chromosome Location
-
Chr.1: 24643012 - 24673281 on GRCh38
- UniGene ID
- Hs.18192
- Species
-
Human