gRNA Design Details
- Target DNA Sequence
- GCGCAAACTTTTGCAGCTGT
- PAM Sequence
- CGG
- Target locus
-
Chr.11: 104163871 - 104163893 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.2/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PDGFD
- Entrez Gene ID
-
80310
- Gene Name
- platelet derived growth factor D
- Gene Aliases
-
IEGF, MSTP036, SCDGF-B, SCDGFB
- Chromosome Location
-
Chr.11: 103907186 - 104164299 on GRCh38
- UniGene ID
- Hs.352298
- Species
-
Human