gRNA Design Details
- Target DNA Sequence
- GGAGATCCGCTCTGTACACG
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 643242 - 643264 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
94.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF22
- Entrez Gene ID
-
27006
- Gene Name
- fibroblast growth factor 22
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 639895 - 643703 on GRCh38
- UniGene ID
- Hs.248087
- Species
-
Human