gRNA Design Details
- Target DNA Sequence
- CACGCGCAGGAAGAAGTGAG
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 640068 - 640090 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.67/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF22
- Entrez Gene ID
-
27006
- Gene Name
- fibroblast growth factor 22
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 639895 - 643703 on GRCh38
- UniGene ID
- Hs.248087
- Species
-
Human