gRNA Design Details
- Target DNA Sequence
- GCAGACCAGGTTGGACATCT
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 33246653 - 33246675 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.71/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK4
- Entrez Gene ID
-
27290
- Gene Name
- serine peptidase inhibitor, Kazal type 4
- Gene Aliases
-
HEL136, PEC-60, PEC60
- Chromosome Location
-
Chr.9: 33240198 - 33248567 on GRCh38
- UniGene ID
- Hs.555934
- Species
-
Human