gRNA Design Details
- Target DNA Sequence
- CGTACCCGTTTTCCAGCGTC
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 4370726 - 4370748 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF23
- Entrez Gene ID
-
8074
- Gene Name
- fibroblast growth factor 23
- Gene Aliases
-
ADHR, FGFN, HPDR2, HYPF, PHPTC
- Chromosome Location
-
Chr.12: 4368227 - 4379728 on GRCh38
- UniGene ID
- Hs.287370
- Species
-
Human