gRNA Design Details

Target DNA Sequence
ACCCTCTTCCACCACCTTTC
PAM Sequence
AGG
Target locus
Chr.9: 120714007 - 120714029 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 83.75/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MEGF9
Entrez Gene ID
1955
Gene Name
multiple EGF like domains 9
Gene Aliases
EGFL5
Chromosome Location
Chr.9: 120600918 - 120714487 on GRCh38
UniGene ID
Hs.744903
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001080497.2 NP_001073966.2 1