gRNA Design Details
- Target DNA Sequence
- GCAAACGAATATGATTCCAA
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 106079514 - 106079536 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.43/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AASDHPPT
- Entrez Gene ID
-
60496
- Gene Name
- aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
- Gene Aliases
-
AASD-PPT, ACPS, CGI-80, LYS2, LYS5
- Chromosome Location
-
Chr.11: 106077565 - 106098692 on GRCh38
- UniGene ID
- Hs.524009
- Species
-
Human