gRNA Design Details
- Target DNA Sequence
- AGGATTAGGCCGCTTCACCA
- PAM Sequence
- GGG
- Target locus
-
Chr.16: 2756507 - 2756529 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.11/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRRM2
- Entrez Gene ID
-
23524
- Gene Name
- serine/arginine repetitive matrix 2
- Gene Aliases
-
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300
- Chromosome Location
-
Chr.16: 2752329 - 2771412 on GRCh38
- UniGene ID
- Hs.433343
- Species
-
Human