gRNA Design Details

Target DNA Sequence
TAGCGATCTCCTAGATAACG
PAM Sequence
TGG
Target locus
Chr.1: 160637319 - 160637341 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 93.59/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SLAMF1
Entrez Gene ID
6504
Gene Name
signaling lymphocytic activation molecule family member 1
Gene Aliases
CD150, CDw150, SLAM
Chromosome Location
Chr.1: 160608103 - 160647537 on GRCh38
UniGene ID
Hs.523660
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_003037.3 NP_003028.1 2
NR_104399.1 2
NR_104400.1 2
NR_104401.1 2
XM_005245456.3 XP_005245513.1 2
XM_011509905.2 XP_011508207.1 2
XM_017002129.1 XP_016857618.1 2
XM_017002130.1 XP_016857619.1 2
XM_017002131.1 XP_016857620.1 2