gRNA Design Details

Target DNA Sequence
GATCCTTCGGGCCATTGGGA
PAM Sequence
AGG
Target locus
Chr.10: 132245914 - 132245936 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.24/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
STK32C
Entrez Gene ID
282974
Gene Name
serine/threonine kinase 32C
Gene Aliases
PKE, YANK3
Chromosome Location
Chr.10: 132207482 - 132332559 on GRCh38
UniGene ID
Hs.469002
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001318878.1 NP_001305807.1 2
NM_001318879.1 NP_001305808.1 2
NM_173575.3 NP_775846.2 2
NR_134911.1 2
XM_011539688.1 XP_011537990.1 2
XM_011539690.2 XP_011537992.1 2
XM_011539693.2 XP_011537995.1 2
XM_011539694.1 XP_011537996.1 2
XM_011539695.1 XP_011537997.1 2
XM_011539696.1 XP_011537998.1 2
XM_017016101.1 XP_016871590.1 2