gRNA Design Details
- Target DNA Sequence
- ACTATATTAGGGCGCTGGTG
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 156810958 - 156810980 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PDGFC
- Entrez Gene ID
-
56034
- Gene Name
- platelet derived growth factor C
- Gene Aliases
-
FALLOTEIN, SCDGF
- Chromosome Location
-
Chr.4: 156761611 - 156971394 on GRCh38
- UniGene ID
- Hs.570855
- Species
-
Human