gRNA Design Details

Target DNA Sequence
ACTATATTAGGGCGCTGGTG
PAM Sequence
TGG
Target locus
Chr.4: 156810958 - 156810980 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 90.49/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PDGFC
Entrez Gene ID
56034
Gene Name
platelet derived growth factor C
Gene Aliases
FALLOTEIN, SCDGF
Chromosome Location
Chr.4: 156761611 - 156971394 on GRCh38
UniGene ID
Hs.570855
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_016205.2 NP_057289.1 3
NR_036641.1 4
XM_011532124.2 XP_011530426.1 5
XM_017008455.1 XP_016863944.1 3
XM_017008456.1 XP_016863945.1 3