gRNA Design Details
- Target DNA Sequence
- TGTGATGACAGCAACGCTCC
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 129800671 - 129800693 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
82.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk35
- Entrez Gene ID
-
67333
- Gene Name
- serine/threonine kinase 35
- Gene Aliases
-
1700054C12Rik, CLIK1, Stk35l1
- Chromosome Location
-
Chr.2: 129800517 - 129832287 on GRCm38
- UniGene ID
- Mm.389329
- Species
-
Mouse