gRNA Design Details
- Target DNA Sequence
- TGTTGCCGTGACTCATGCGC
- PAM Sequence
- TGG
- Target locus
-
Chr.20: 2103298 - 2103320 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK35
- Entrez Gene ID
-
140901
- Gene Name
- serine/threonine kinase 35
- Gene Aliases
-
CLIK1, STK35L1
- Chromosome Location
-
Chr.20: 2098035 - 2148555 on GRCh38
- UniGene ID
- Hs.100057
- Species
-
Human