gRNA Design Details

Target DNA Sequence
GACTTCACAGGTCAGGGTTA
PAM Sequence
CGG
Target locus
Chr.6: 32182607 - 32182629 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 94.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AGER
Entrez Gene ID
177
Gene Name
advanced glycosylation end product-specific receptor
Gene Aliases
RAGE, SCARJ1
Chromosome Location
Chr.6: 32180968 - 32185247 on GRCh38
UniGene ID
Hs.534342
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001136.4 NP_001127.1 7
NM_001206929.1 NP_001193858.1 7
NM_001206932.1 NP_001193861.1 7
NM_001206934.1 NP_001193863.1 7
NM_001206936.1 NP_001193865.1 7
NM_001206940.1 NP_001193869.1 7
NM_001206954.1 NP_001193883.1 7
NM_001206966.1 NP_001193895.1 7
NM_172197.2 NP_751947.1 7
NR_038190.1 6
XM_017010328.1 XP_016865817.1 7