gRNA Design Details
- Target DNA Sequence
- GTGTATGCAGCATTCTGCCG
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 58285987 - 58286009 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS54
- Entrez Gene ID
-
221191
- Gene Name
- protease, serine 54
- Gene Aliases
-
CT67, KLKBL4
- Chromosome Location
-
Chr.16: 58279997 - 58295047 on GRCh38
- UniGene ID
- Hs.411239
- Species
-
Human