gRNA Design Details
- Target DNA Sequence
- ATGCGGCAGCCACACGAAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 58284610 - 58284632 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS54
- Entrez Gene ID
-
221191
- Gene Name
- protease, serine 54
- Gene Aliases
-
CT67, KLKBL4
- Chromosome Location
-
Chr.16: 58279997 - 58295047 on GRCh38
- UniGene ID
- Hs.411239
- Species
-
Human