gRNA Design Details
- Target DNA Sequence
- TTCAGGCAACGACATTTCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.6: 44360496 - 44360518 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.57/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPATS1
- Entrez Gene ID
-
221409
- Gene Name
- spermatogenesis associated serine rich 1
- Gene Aliases
-
DDIP, SPATA8, SRSP1
- Chromosome Location
-
Chr.6: 44342660 - 44377167 on GRCh38
- UniGene ID
- Hs.135283
- Species
-
Human