gRNA Design Details
- Target DNA Sequence
- AATATAGCTGTCCAAGTGCG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 38413274 - 38413296 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.83/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBPL1
- Entrez Gene ID
-
347252
- Gene Name
- insulin like growth factor binding protein like 1
- Gene Aliases
-
IGFBP-RP4, IGFBPRP4, bA113O24.1
- Chromosome Location
-
Chr.9: 38406528 - 38424447 on GRCh38
- UniGene ID
- Hs.349705
- Species
-
Human