gRNA Design Details
- Target DNA Sequence
- AATGGCGATGACCACTCAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 216660559 - 216660581 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
94.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP2
- Entrez Gene ID
-
3485
- Gene Name
- insulin like growth factor binding protein 2
- Gene Aliases
-
IBP2, IGF-BP53
- Chromosome Location
-
Chr.2: 216632828 - 216664436 on GRCh38
- UniGene ID
- Hs.438102
- Species
-
Human