gRNA Design Details
- Target DNA Sequence
- AGTGACCTTCTCCCGGAACA
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 216660680 - 216660702 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP2
- Entrez Gene ID
-
3485
- Gene Name
- insulin like growth factor binding protein 2
- Gene Aliases
-
IBP2, IGF-BP53
- Chromosome Location
-
Chr.2: 216632828 - 216664436 on GRCh38
- UniGene ID
- Hs.438102
- Species
-
Human