gRNA Design Details

Target DNA Sequence
AGTGACCTTCTCCCGGAACA
PAM Sequence
CGG
Target locus
Chr.2: 216660680 - 216660702 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.68/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGFBP2
Entrez Gene ID
3485
Gene Name
insulin like growth factor binding protein 2
Gene Aliases
IBP2, IGF-BP53
Chromosome Location
Chr.2: 216632828 - 216664436 on GRCh38
UniGene ID
Hs.438102
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000597.2 NP_000588.2 2
NM_001313990.1 NP_001300919.1 2
NM_001313992.1 NP_001300921.1 2
NM_001313993.1 NP_001300922.1 2