gRNA Design Details
- Target DNA Sequence
- GCAGTATAGACACTCGTCAC
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 118933082 - 118933104 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TNFRSF11B
- Entrez Gene ID
-
4982
- Gene Name
- TNF receptor superfamily member 11b
- Gene Aliases
-
OCIF, OPG, PDB5, TR1
- Chromosome Location
-
Chr.8: 118923557 - 118952144 on GRCh38
- UniGene ID
- Hs.81791
- Species
-
Human